Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145098.2 | NP_659566.1 | 5 | 1005 | ||
XM_006255825.2 | XP_006255887.1 | 5 | 1198 | |||
XM_006255826.2 | XP_006255888.1 | 4 | 983 | |||
XM_006255827.2 | XP_006255889.1 | 4 | 983 | |||
XM_008772598.1 | XP_008770820.1 | 4 | 983 | |||
GenBank mRNA | AF016296.1 | - | 4 | 609 | ||
AF018957.1 | - | 4 | 462 | |||
BC085689.1 | - | 5 | 1005 | 7109697 |