Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other THBS3 SIRNA Assays ›
Gene Symbol:
THBS3
Gene Name:
thrombospondin 3
Gene Aliases:
TSP3
Chromosome Location:
Chr.1: 155195588 - 155209180 on Build GRCh38
UniGene ID:
Hs.169875
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s14104
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: THBS3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001252607.1
NP_001239536.1
12, 13
1618
NM_001252608.1
NP_001239537.1
11, 12
1285
NM_007112.4
NP_009043.1
13, 14
1645
NR_045553.1
12, 13
1629
XM_006711498.3
XP_006711561.1
13, 14
3204
XM_011509926.2
XP_011508228.1
13, 14
3204
XM_011509927.2
XP_011508229.1
12, 13
3177
XM_011509928.2
XP_011508230.1
13, 14
3111
XM_011509929.1
XP_011508231.1
13, 14
1600
XM_011509930.2
XP_011508232.1
13, 14
3204
XM_011509931.1
XP_011508233.1
13, 14
1550
XM_011509932.2
XP_011508234.1
13, 14
1483
XM_011509933.1
XP_011508235.1
14, 15
1832
XM_011509934.1
XP_011508236.1
13, 14
1587
XM_011509935.1
XP_011508237.1
13, 14
1743
XM_011509936.1
XP_011508238.1
12, 13
1484
XM_017002206.1
XP_016857695.1
12, 13
1557
GenBank mRNA
AK074796.1
-
10, 11
981
AK223278.1
-
13, 14
1559
AK298592.1
-
11, 12
1221
AK298914.1
-
3, 4
295
AK314337.1
-
12, 13
1569
BC018786.1
-
4, 5
448
BC113846.1
-
12, 13
1547
40008346
BC113847.1
-
13, 14
1563
40008343
L38969.1
-
13, 14
1561
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
growth plate cartilage development
cell-matrix adhesion
ossification involved in bone maturation
bone trabecula formation
calcium ion binding
heparin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0