Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THBS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252607.1 | NP_001239536.1 | 10 | 1370 | ||
NM_001252608.1 | NP_001239537.1 | 9 | 1037 | |||
NM_007112.4 | NP_009043.1 | 11 | 1397 | |||
NR_045553.1 | 10 | 1381 | ||||
XM_006711498.3 | XP_006711561.1 | 11 | 2956 | |||
XM_011509926.2 | XP_011508228.1 | 11 | 2956 | |||
XM_011509927.2 | XP_011508229.1 | 10 | 2929 | |||
XM_011509928.2 | XP_011508230.1 | 11 | 2863 | |||
XM_011509929.1 | XP_011508231.1 | 11 | 1352 | |||
XM_011509930.2 | XP_011508232.1 | 11 | 2956 | |||
XM_011509931.1 | XP_011508233.1 | 11 | 1302 | |||
XM_011509932.2 | XP_011508234.1 | 11 | 1235 | |||
XM_011509933.1 | XP_011508235.1 | 12 | 1584 | |||
XM_011509934.1 | XP_011508236.1 | 11 | 1339 | |||
XM_011509935.1 | XP_011508237.1 | 11 | 1495 | |||
XM_011509936.1 | XP_011508238.1 | 10 | 1236 | |||
XM_017002206.1 | XP_016857695.1 | 10 | 1309 | |||
GenBank mRNA | AK074796.1 | - | 8 | 733 | ||
AK223278.1 | - | 11 | 1311 | |||
AK298592.1 | - | 9 | 973 | |||
AK298803.1 | - | 8 | 865 | |||
AK298914.1 | - | 2 | 158 | |||
AK314337.1 | - | 10 | 1321 | |||
BC018786.1 | - | 2 | 200 | |||
BC113846.1 | - | 10 | 1299 | 40008346 | ||
BC113847.1 | - | 11 | 1315 | 40008343 | ||
L38969.1 | - | 11 | 1313 |