Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gimap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033913.1 | NP_001029085.1 | 3 | 480 | ||
NM_145680.3 | NP_663713.1 | 2, 3 | 305 | |||
XM_006236457.2 | XP_006236519.1 | 3 | 843 | |||
XM_006236458.2 | XP_006236520.1 | 2, 3 | 651 | |||
XM_008762871.1 | XP_008761093.1 | 3, 4 | 710 | |||
GenBank mRNA | AY055776.3 | - | 2, 3 | 305 | ||
AY055777.2 | - | 1 | 192 | |||
AY550018.1 | - | 3 | 480 | |||
AY550021.1 | - | 5 | 998 | |||
AY550022.1 | - | 4, 5 | 823 | |||
AY550023.1 | - | 3, 4 | 715 | |||
BC092561.1 | - | 3 | 334 | 7130634 | ||
DQ125352.1 | - | 2, 3 | 136 | |||
DQ125353.1 | - | 2, 3 | 136 |