Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025271.1 | NP_001020442.1 | 9 | 1973 | ||
GenBank mRNA | AF036335.1 | - | 5 | 503 | ||
BC098731.1 | - | 9 | 1973 | 7384804 | ||
FQ227268.1 | - | 5 | 438 | |||
FQ229853.1 | - | 5 | 515 |