Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761973.1 | XP_008760195.1 | 14 | 1394 | ||
GenBank mRNA | AB031229.1 | - | 14 | 1570 |