Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024964.1 | NP_001020135.1 | 4 | 754 | ||
XM_006227767.2 | XP_006227829.1 | 4 | 872 | |||
XM_006227768.2 | XP_006227830.1 | 4 | 1168 | |||
GenBank mRNA | BC097993.1 | - | 4 | 754 | 7446619 | |
U32575.1 | - | 3 | 709 |