Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032282.3 | NP_001027453.1 | 3 | 1030 | ||
NM_005655.3 | NP_005646.1 | 3 | 872 | |||
GenBank mRNA | AK312894.1 | - | 3 | 638 | ||
BC011538.1 | - | 3 | 857 | 3868209 | ||
BC095399.1 | - | 3 | 693 | 30520399 | ||
BT006634.1 | - | 2 | 538 | |||
S81439.1 | - | 3 | 1006 | |||
U21847.1 | - | 3 | 657 |