Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147210.2 | NP_671743.2 | 5 | 915 | ||
GenBank mRNA | U15660.1 | - | 4 | 631 | ||
U20796.1 | - | 4 | 572 | |||
X78135.1 | - | 5 | 768 | |||
X82777.1 | - | 5 | 850 |