Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181365.2 | NP_852030.1 | 658 | |||
GenBank mRNA | AF345444.1 | - | 658 | |||
AF345445.1 | - | 1, 2 | 228 | |||
AF453245.1 | - | 2, 3 | 281 |