Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148890.1 | NP_683688.1 | 2, 3 | 260 | ||
GenBank mRNA | AY043393.1 | - | 2, 3 | 260 |