Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148891.2 | NP_683689.1 | 3 | 650 | ||
GenBank mRNA | AJ492222.1 | - | 3 | 271 | ||
BC097277.1 | - | 3 | 650 | 7457194 |