Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152242.2 | NP_689448.2 | 9 | 1391 | ||
XM_006255066.2 | XP_006255128.1 | 9 | 2443 | |||
XM_008772264.1 | XP_008770486.1 | 9 | 1593 | |||
XM_008772265.1 | XP_008770487.1 | 10 | 1857 | |||
XM_008772266.1 | XP_008770488.1 | 9 | 3103 | |||
XM_008772267.1 | XP_008770489.1 | 9 | 3366 | |||
XM_008772268.1 | XP_008770490.1 | 10 | 1690 | |||
XM_008772269.1 | XP_008770491.1 | 9 | 2472 | |||
XM_008772270.1 | XP_008770492.1 | 9 | 2948 | |||
GenBank mRNA | AF529886.1 | - | 9 | 1392 | ||
EX488169.1 | - | 6 | 502 |
Set Membership: |
G-protein coupled receptor |