Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253357.1 | NP_001240286.1 | 8 | 1183 | ||
NM_005424.4 | NP_005415.1 | 8 | 1266 | |||
XM_005271163.1 | XP_005271220.1 | 7 | 1080 | |||
XM_006710869.1 | XP_006710932.1 | 8 | 1400 | |||
XM_017002207.1 | XP_016857696.1 | 8 | 1209 | |||
GenBank mRNA | AK225560.1 | - | 8 | 1144 | ||
AK296735.1 | - | 3 | 242 | |||
AK300399.1 | - | 8 | 1073 | |||
AK300623.1 | - | 8 | 966 | |||
BC038239.1 | - | 8 | 1168 | 5767075 | ||
EU826590.1 | - | 8 | 1089 | |||
X60957.1 | - | 8 | 1125 |