Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153300.1 | NP_695212.1 | 7, 8 | 778 | ||
GenBank mRNA | AB369261.1 | - | 7, 8 | 778 | ||
AF434845.1 | - | 7, 8 | 778 | |||
BC166415.1 | - | 835 | 9025597 |