Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253357.1 | NP_001240286.1 | 16, 17 | 2826 | ||
NM_005424.4 | NP_005415.1 | 16, 17 | 2909 | |||
XM_005271163.1 | XP_005271220.1 | 15, 16 | 2723 | |||
XM_006710869.1 | XP_006710932.1 | 16, 17 | 3043 | |||
GenBank mRNA | AF217976.1 | - | 6, 7 | 993 | ||
AK225560.1 | - | 16, 17 | 2568 | |||
AK296735.1 | - | 11, 12 | 1885 | |||
AK300399.1 | - | 16, 17 | 2716 | |||
AK307639.1 | - | 9, 10 | 1577 | |||
BC038239.1 | - | 16, 17 | 2811 | 5767075 | ||
X60957.1 | - | 16, 17 | 2768 |