Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085381.1 | NP_001078850.1 | 1559 | |||
GenBank mRNA | BC166405.1 | - | 1444 | 9021864 |