Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270871.1 | NP_001257800.1 | 22, 23 | 2323 | ||
NM_001270872.1 | NP_001257801.1 | 21, 22 | 2287 | |||
NM_181366.2 | NP_852031.1 | 23, 24 | 2371 | |||
XM_006256916.2 | XP_006256978.1 | 23, 24 | 2411 | |||
XM_008773154.1 | XP_008771376.1 | 22, 23 | 2751 | |||
XM_008773155.1 | XP_008771377.1 | 22, 23 | 2361 | |||
XM_008773156.1 | XP_008771378.1 | 21, 22 | 2715 | |||
XM_008773157.1 | XP_008771379.1 | 21, 22 | 2709 | |||
XM_008773158.1 | XP_008771380.1 | 20, 21 | 2673 | |||
XM_008773159.1 | XP_008771381.1 | 19, 20 | 2264 | |||
XM_008773160.1 | XP_008771382.1 | 22, 23 | 2752 | |||
GenBank mRNA | AF538953.1 | - | 23, 24 | 2381 | ||
AF538958.1 | - | 22, 23 | 2333 | |||
AF538959.1 | - | 21, 22 | 2297 |
Set Membership: |
G-protein coupled receptor |