Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153627.1 | NP_705891.1 | 2 | 268 | ||
XM_008767664.1 | XP_008765886.1 | 2 | 265 | |||
GenBank mRNA | AB037922.1 | - | 2 | 268 |