Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153629.1 | NP_705893.1 | 11 | 1583 | ||
XM_006246232.1 | XP_006246294.1 | 11 | 1627 | |||
GenBank mRNA | AF077354.1 | - | 11 | 1583 | ||
FQ222433.1 | - | 12 | 1791 |