Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172047.1 | NP_742044.1 | 5 | 712 | ||
XM_008768707.1 | XP_008766929.1 | 5 | 599 | |||
XM_008768708.1 | XP_008766930.1 | 3 | 1268 | |||
XM_008768709.1 | XP_008766931.1 | 3 | 998 | |||
GenBank mRNA | AY049022.1 | - | 5 | 712 |