Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007606.1 | NP_001007607.1 | 4, 5 | 513 | ||
XM_006233136.2 | XP_006233198.1 | 4, 5 | 548 | |||
GenBank mRNA | BC061765.1 | - | 4, 5 | 513 | 5620893 |