Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172032.2 | NP_742029.3 | 2 | 251 | ||
GenBank mRNA | AF508022.1 | - | 382 | |||
BC060541.1 | - | 2 | 241 | |||
BC088106.1 | - | 2 | 251 | 7380129 |