Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236922.2 | XP_006236984.1 | 18 | 3630 | ||
XM_006236923.2 | XP_006236985.1 | 18 | 3442 | |||
XM_006236924.2 | XP_006236986.1 | 17 | 3489 | |||
XM_008763139.1 | XP_008761361.1 | 18 | 3376 |