Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrfap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009264.1 | NP_001009264.1 | 1 | 299 | ||
GenBank mRNA | AF526268.1 | - | 2 | 249 | ||
BC088308.1 | - | 1 | 299 | 7324285 | ||
FQ228750.1 | - | 1 | 319 |