Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232412.2 | XP_006232474.1 | 8, 9 | 2153 | ||
XM_006232430.2 | XP_006232492.1 | 7, 8 | 1495 | |||
XM_006232434.2 | XP_006232496.1 | 5, 6 | 962 | |||
XM_008761033.1 | XP_008759255.1 | 6, 7 | 1093 | |||
XM_008761035.1 | XP_008759257.1 | 6, 7 | 1036 | |||
XM_008761036.1 | XP_008759258.1 | 7, 8 | 1147 | |||
XM_008761038.1 | XP_008759260.1 | 5, 6 | 982 | |||
XM_008761043.1 | XP_008759265.1 | 5, 6 | 982 |