Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wrnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172332.2 | NP_758835.1 | 6 | 1868 | ||
GenBank mRNA | AY136827.1 | - | 6 | 1772 | ||
BC098652.1 | - | 6 | 1868 | 7306780 |