Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fibp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172334.2 | NP_758837.1 | 1 | 68 | ||
XM_006230701.2 | XP_006230763.1 | 1 | 124 | |||
GenBank mRNA | AY093421.1 | - | 1 | 69 | ||
BC061802.1 | - | 1 | 89 | 5598566 | ||
FQ228181.1 | - | 1 | 120 | |||
FQ234469.1 | - | 1 | 120 | |||
FQ234564.1 | - | 1 | 120 |