Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204760.1 | NP_001191689.1 | 6 | 1425 | ||
NM_012464.4 | NP_036596.3 | 6 | 1425 | |||
XM_011532214.1 | XP_011530516.1 | 6 | 1483 | |||
XM_017008570.1 | XP_016864059.1 | 7 | 1551 | |||
GenBank mRNA | AF282732.1 | - | 6 | 1029 | ||
BC016922.2 | - | 6 | 943 | |||
BC136429.1 | - | 6 | 1335 | 9020418 | ||
BC136430.1 | - | 6 | 1335 | 9020419 | ||
BC144085.1 | - | 6 | 1335 | 9052605 | ||
U91963.1 | - | 6 | 1425 |