Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134350.2 | NP_599177.2 | 10 | 1264 | ||
XM_006248150.2 | XP_006248212.1 | 12 | 1408 | |||
XM_008768568.1 | XP_008766790.1 | 12 | 1342 | |||
GenBank mRNA | BC099808.1 | - | 10 | 1264 | 7938638 | |
X52712.1 | - | 10 | 1339 | |||
X52713.1 | - | 9 | 1144 |