Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006289.3 | NP_006280.3 | 10, 11 | 1466 | ||
GenBank mRNA | AB028950.2 | - | 10, 11 | 1441 | ||
AF078828.1 | - | 10, 11 | 1238 | |||
AF177198.1 | - | 10, 11 | 1202 | |||
BC042923.1 | - | 10, 11 | 1252 | 5576203 |