Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugt2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173295.1 | NP_775417.1 | 5 | 1235 | ||
GenBank mRNA | M13506.1 | - | 5 | 1235 |