Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1271 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173300.1 | NP_775422.1 | 2 | 977 | ||
GenBank mRNA | X80671.1 | - | 2 | 977 |
Gene Symbol: LOC100909885 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749179.2 | XP_003749227.2 | 1 | 772 |
Gene Symbol: LOC100910120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750489.2 | XP_003750537.2 | 1 | 769 |
Gene Symbol: LOC100910162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749263.2 | XP_003749311.2 | 772 |
Gene Symbol: LOC100910224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008766109.1 | XP_008764331.1 | 1 | 772 |
Gene Symbol: LOC100910294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232223.1 | XP_006232285.1 | 1 | 772 |