Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173302.1 | NP_775424.1 | 1384 | |||
GenBank mRNA | AB051836.1 | - | 1384 |