Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2d5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173304.2 | NP_775426.1 | 6 | 985 | ||
GenBank mRNA | BC088097.1 | - | 6 | 987 | 7369436 | |
J02869.1 | - | 6 | 985 | |||
M22329.1 | - | 6 | 919 |
Gene Symbol: Cyp2d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153313.1 | NP_695225.1 | 6 | 1020 | ||
GenBank mRNA | AB008422.1 | - | 6 | 1020 | ||
BC078696.1 | - | 6 | 1027 | 7126263 | ||
FQ219613.1 | - | 6 | 1070 | |||
J02867.1 | - | 6 | 1020 | |||
M16654.1 | - | 6 | 1020 | |||
M22328.1 | - | 6 | 1036 |