Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173317.1 | NP_775439.1 | 2673 | |||
GenBank mRNA | AF016180.1 | - | 2673 |
Gene Symbol: Vom2r41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099500.1 | NP_001092970.1 | 6 | 2033 |
Gene Symbol: Vom2r42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099511.1 | NP_001092981.1 | 6 | 2051 |
Set Membership: |
G-protein coupled receptor |