Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001638.2 | NP_001629.1 | 2 | 109 | ||
GenBank mRNA | AK314892.1 | - | 2 | 100 | ||
BC026257.1 | - | 2 | 109 | 4710467 | ||
CR936663.1 | - | 2 | 134 | |||
L27050.1 | - | 2 | 105 |