Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1361 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173333.1 | NP_775455.1 | 1 | 798 | ||
GenBank mRNA | M64377.1 | - | 1 | 798 |
Gene Symbol: LOC100912159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750781.2 | XP_003750829.2 | 2 | 839 |
Gene Symbol: Olr1358 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214822.1 | NP_999987.1 | 1 | 798 |
Gene Symbol: Olr1365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214824.1 | NP_999989.1 | 798 |