Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1361 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173333.1 | NP_775455.1 | 1 | 470 | ||
GenBank mRNA | M64377.1 | - | 1 | 470 |
Gene Symbol: Olr1358 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214822.1 | NP_999987.1 | 1 | 470 |
Gene Symbol: Olr1362 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006598.1 | NP_001006598.1 | 1 | 470 | ||
GenBank mRNA | AF091566.1 | - | 1 | 281 |