Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214823.1 | NP_999988.1 | 1 | 266 |
Gene Symbol: Olr1525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000529.1 | NP_001000529.1 | 266 | |||
XM_008767530.1 | XP_008765752.1 | 1 | 266 |