Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214825.1 | NP_999990.1 | 1 | 470 |
Gene Symbol: LOC100912159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750781.2 | XP_003750829.2 | 2 | 511 |
Gene Symbol: Olr1365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214824.1 | NP_999989.1 | 470 |