Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kremen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105767.1 | NP_001099237.1 | 5 | 575 | ||
XM_008767550.1 | XP_008765772.1 | 4 | 798 | |||
XM_008767551.1 | XP_008765773.1 | 4 | 798 | |||
XM_008767552.1 | XP_008765774.1 | 4 | 798 |