Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199371.1 | NP_955403.1 | 5 | 662 | ||
XM_006245922.2 | XP_006245984.1 | 6 | 853 | |||
XM_006245923.2 | XP_006245985.1 | 6 | 861 | |||
XM_008767553.1 | XP_008765775.1 | 6 | 881 | |||
GenBank mRNA | AB073023.1 | - | 5 | 662 |