Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009620.1 | NP_001009620.1 | 1, 2 | 824 | ||
GenBank mRNA | BC087601.1 | - | 1, 2 | 824 | 7314703 |