Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR137B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003272.3 | NP_003263.1 | 5 | 1013 | ||
XM_017002209.1 | XP_016857698.1 | 5 | 1043 | |||
GenBank mRNA | AF027826.1 | - | 5 | 973 | ||
AK223632.1 | - | 5 | 992 | |||
AK295408.1 | - | 6 | 925 | |||
AK301552.1 | - | 6 | 945 | |||
AK312908.1 | - | 5 | 1006 | |||
AY242135.1 | - | 5 | 922 | |||
BC012100.1 | - | 5 | 1013 | 4643216 | ||
CR541770.1 | - | 5 | 922 |