Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007607.1 | NP_001007608.1 | 2 | 430 | ||
XM_006245942.2 | XP_006246004.1 | 2 | 509 | |||
XM_008767559.1 | XP_008765781.1 | 2 | 449 | |||
XM_008767560.1 | XP_008765782.1 | 2 | 476 | |||
GenBank mRNA | BC081875.1 | - | 2 | 430 | 7107681 |