Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC287167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013853.1 | NP_001013875.1 | 1, 2 | 100 | ||
GenBank mRNA | FQ211031.1 | - | 1, 2 | 131 | ||
FQ212394.1 | - | 1, 2 | 131 | |||
FQ218263.1 | - | 1, 2 | 131 | |||
FQ224052.1 | - | 1, 2 | 131 | |||
FQ226501.1 | - | 1, 2 | 133 | |||
FQ228302.1 | - | 1, 2 | 131 |