Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105777.2 | NP_001099247.2 | 6 | 667 | ||
GenBank mRNA | BC161929.1 | - | 6 | 653 | 8375237 | |
CF110623.1 | - | 2 | 146 |