Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007114.2 | NP_009045.2 | 2 | 925 | ||
XM_011534056.2 | XP_011532358.1 | 2 | 989 | |||
GenBank mRNA | AB209109.1 | - | 2 | 895 | ||
AK299020.1 | - | 3 | 576 | |||
AK312697.1 | - | 2 | 879 | |||
BC033771.2 | - | 2 | 870 | |||
BC117418.1 | - | 2 | 778 | 40125969 | ||
BC126123.1 | - | 2 | 778 | 8991839 | ||
BC143840.1 | - | 2 | 778 | 9052355 | ||
L01042.1 | - | 2 | 678 |