Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007608.2 | NP_001007609.1 | 2 | 156 | ||
XM_006246244.2 | XP_006246306.1 | 2 | 455 | |||
GenBank mRNA | BC058152.1 | - | 2 | 159 | 6890712 | |
FQ210698.1 | - | 1 | 54 | |||
FQ212502.1 | - | 1 | 54 | |||
FQ212528.1 | - | 1 | 54 | |||
FQ212745.1 | - | 1 | 54 | |||
FQ212916.1 | - | 1 | 54 | |||
FQ213991.1 | - | 1 | 54 | |||
FQ214002.1 | - | 1 | 54 | |||
FQ214005.1 | - | 1 | 54 | |||
FQ214369.1 | - | 1 | 54 | |||
FQ214470.1 | - | 1 | 54 | |||
FQ214982.1 | - | 1 | 54 | |||
FQ215334.1 | - | 1 | 54 | |||
FQ215452.1 | - | 1 | 54 | |||
FQ219088.1 | - | 1 | 54 | |||
FQ219401.1 | - | 1 | 54 | |||
FQ224225.1 | - | 1 | 54 | |||
FQ224935.1 | - | 1 | 54 | |||
FQ229076.1 | - | 1 | 54 | |||
FQ229788.1 | - | 54 | ||||
FQ230985.1 | - | 1 | 54 |